Invitrogen
Manufacturer Code:MA536238
Catalog # PIMA536238
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 11I4 |
Immunogen | A synthetic peptide corresponding to a sequence of human ASXL1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: additional sex combs like 1; additional sex combs like 1 additional sex combs like 1 transcriptional regulator additional sex combs like transcriptional regulator 1 additional sex combs-like protein 1 Asxl1 BOPS KIAA0978 MDS MGC117280 MGC71111 mKIAA0978 Polycomb group protein ASXL1 putative Polycomb group protein ASXL1 RGD1561878; additional sex combs like transcriptional regulator 1; Additional sex combs-like protein 1; Polycomb group protein ASXL1; putative Polycomb group protein ASXL1
Gene Aliases: ASXL1; BOPS; KIAA0978; MDS; mKIAA0978; RGD1561878
UniProt ID: (Human) Q8IXJ9, (Mouse) P59598
Entrez Gene ID: (Human) 171023, (Mouse) 228790, (Rat) 311553
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.