Invitrogen
Manufacturer Code:PA568360
Catalog # PIPA568360
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human ASXL1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: additional sex combs like 1; additional sex combs like transcriptional regulator 1; Additional sex combs-like protein 1; KIAA0978; Polycomb group protein ASXL1; putative Polycomb group protein ASXL1
Gene Aliases: ASXL1; BOPS; KIAA0978; MDS; mKIAA0978
UniProt ID: (Human) Q8IXJ9, (Mouse) P59598
Entrez Gene ID: (Human) 171023, (Mouse) 228790
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.