Novus Biologicals
Manufacturer Code:NBP29259020UL
Catalog # NB027051
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 70-250 of human ASXL1 (NP_056153.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: additional sex combs like 1; additional sex combs like 1 (Drosophila); additional sex combs like transcriptional regulator 1; Additional sex combs-like protein 1; Polycomb group protein ASXL1
Gene Aliases: ASXL1; BOPS; KIAA0978; MDS
UniProt ID: (Human) Q8IXJ9
Entrez Gene ID: (Human) 171023
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.