Proteintech
Manufacturer Code:198921AP150UL
Catalog # 50-173-2588
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ASUN Fusion Protein Ag13727 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ASUN C12orf11 GCT1 NET48 NY SAR 95 Sarcoma antigen NY SAR 95; asunder spermatogenesis regulator; asunder, spermatogenesis regulator homolog (Drosphila); Cell cycle regulator Mat89Bb homolog; Germ cell tumor 1; Integrator complex subunit 13; likely orthologue of H. sapiens chromosome 12 open reading frame 11 (C12orf11); Protein asunder homolog; Sarcoma antigen NY-SAR-95; spermatogenesis associated 30; Spermatogenesis-associated protein 30
Gene Aliases: 4933424B01Rik; ASUN; C12orf11; GCT1; INTS13; Mat89Bb; NET48; SPATA30
UniProt ID: (Human) Q9NVM9, (Mouse) Q8QZV7
Entrez Gene ID: (Human) 55726, (Rat) 690728, (Mouse) 71177
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.