Proteintech
Manufacturer Code:660361IG150UL
Catalog # 50-173-6476
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1D1A11 |
Immunogen | ASS1 Fusion Protein Ag9314 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Argininosuccinate synthase; Argininosuccinate synthase argininosuccinate synthetase 1 ASS ASS1 Citrulline aspartate ligase CTLN1; argininosuccinate synthetase 1; arginosuccinate synthetase 1; Ass1; Citrulline--aspartate ligase; citrulline-aspartate ligase
Gene Aliases: AA408052; ASS; Ass-1; ASS1; ASSA; CTLN1; fold
UniProt ID: (Human) P00966, (Rat) P09034, (Mouse) P16460
Entrez Gene ID: (Pig) 414411, (Human) 445, (Rat) 25698, (Mouse) 11898
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.