Invitrogen
Manufacturer Code:PA518679
Catalog # PIPA518679
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (ENPKNQAPPGLYTKTQD) corresponding to the internal amino acids of ASS1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: arginino succinate synthetase; Argininosuccinate synthase; argininosuccinate synthetase; argininosuccinate synthetase 1; Argininosuccinate synthetase 1 ASS ASS1 CTLN1 citrulline--aspartate ligase; Citrulline--aspartate ligase; citrulline-aspartate ligase; citrullinemia; mutant arginino succinate synthetase
Gene Aliases: ASS; ASS1; CN; CTLN1
UniProt ID: (Human) P00966, (Bovine) P14568
Entrez Gene ID: (Human) 445, (Bovine) 280726
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.