Abnova
Manufacturer Code:MAB15044
Catalog # 89-035-064
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CRASH/1289 |
Immunogen | Recombinant protein corresponding to full length human ASRGL1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ALP; ALP1; asparaginase-like 1 protein; Asparaginase-like protein 1; Beta-aspartyl-peptidase; FLJ22316; Isoaspartyl dipeptidase; Isoaspartyl peptidase/L-asparaginase; Isoaspartyl peptidase/L-asparaginase alpha chain; Isoaspartyl peptidase/L-asparaginase beta chain; L-asparaginase; L-asparagine amidohydrolase; testis secretory sperm-binding protein Li 242mP
Gene Aliases: ALP; ALP1; ASRGL1; CRASH
UniProt ID: (Human) Q7L266
Entrez Gene ID: (Human) 80150
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.