Invitrogen
Manufacturer Code:PA567689
Catalog # PIPA567689
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human ASPM. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Abnormal spindle protein homolog; Abnormal spindle-like microcephaly-associated protein; asp (abnormal spindle) homolog, microcephaly associated; Asp homolog; MCPH5
Gene Aliases: ASP; ASPM; Calmbp1; MCPH5
UniProt ID: (Human) Q8IZT6
Entrez Gene ID: (Human) 259266
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.