Novus Biologicals
Manufacturer Code:NBP292395
Catalog # NB026666
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human ASPM. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Abnormal spindle protein homolog; Abnormal spindle-like microcephaly-associated protein; asp (abnormal spindle) homolog, microcephaly associated; ASP asp (abnormal spindle) homolog microcephaly associated (Drosophila) Asp homolog Calmbp1 FLJ43117 MCPH5DKFZp686N06184 microcephaly associated (Drosophila) microcephaly primary autosomal recessive 5; Asp homolog
Gene Aliases: ASP; ASPM; Calmbp1; MCPH5
UniProt ID: (Human) Q8IZT6
Entrez Gene ID: (Human) 259266
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.