Invitrogen
Manufacturer Code:PA564429
Catalog # PIPA564429
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ASPG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1-alkyl-2-acetylglycerophosphocholine esterase; 60 kDa lysophospholipase; 60 kDa lysophospholipase L-asparaginase L-asparagine amidohydrolase Platelet-activating factor acetylhydrolase PAF acetylhydrolase ASPG C14orf76; 60-kDa-lysophospholipase; asparaginase homolog; L-asparaginase; L-asparagine amidohydrolase; LysoLP; Lysophospholipase-transacylase; PAF acetylhydrolase; Platelet-activating factor acetylhydrolase; wild-type L-asparaginase
Gene Aliases: ASPG; C14orf76; GPA/WT
UniProt ID: (Human) Q86U10
Entrez Gene ID: (Human) 374569
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.