Novus Biologicals
Manufacturer Code:NBP272508FR
Catalog # NBP272508FR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4G8 |
Immunogen | Full length recombinant protein of human ASPDH (NP_001019827) produced in E.coli. |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: aspartate dehydrogenase domain containing Aspartate dehydrogenase domain-containing protein EC 1.4.1.21 putative L-aspartate dehydrogenase; Aspartate dehydrogenase domain-containing protein; Putative L-aspartate dehydrogenase
Gene Aliases: ASPDH
UniProt ID: (Human) A6ND91
Entrez Gene ID: (Human) 554235
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.