Invitrogen
Manufacturer Code:PA557535
Catalog # PIPA557535
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ASNSD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Asparagine synthetase domain-containing protein 1; HCV NS3-transactivated protein 1; Nbla00058 NBLA00058 NS3TP1 HCV NS3-transactivated protein 1 asparagine synthetase domain-containing protein 1
Gene Aliases: ASNSD1; NBLA00058; NS3TP1
UniProt ID: (Human) Q9NWL6
Entrez Gene ID: (Human) 54529
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.