Invitrogen
Manufacturer Code:PA545067
Catalog # PIPA545067
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human ASNA1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: arsA arsenite transporter, ATP-binding, homolog 1; Arsenical pump-driving ATPase; Arsenical resistance ATPase; Arsenite-stimulated ATPase; Arsenite-translocating ATPase; Arsenite-transporting ATPase; ASNA1; ATPase ASNA1; ATPase GET3; Guided entry of tail-anchored proteins factor 3, ATPase; hARSA-I; hASNA-I; Transmembrane domain recognition complex 40 kDa ATPase subunit
Gene Aliases: 1810048H22Rik; ARSA; ARSA-I; ARSA1; ASNA-I; ASNA1; GET3; si:dkey-121b10.2; TRC40; wu:fd05d03; zgc:56540; zgc:86799
UniProt ID: (Human) O43681, (Mouse) O54984, (Bovine) A5PJI5, (Zebrafish) Q6IQE5
Entrez Gene ID: (Human) 439, (Mouse) 56495, (Rat) 288919, (Guinea pig) 100716352, (Dog) 476699, (Rabbit) 100352737, (Bovine) 504586, (Horse) 100063417, (Zebrafish) 325704
Molecular Function:
hydrolase
nucleotide phosphatase
phosphatase
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.