Abnova
Manufacturer Code:PAB11499
Catalog # 89-115-133
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human ASNA1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of ASNA1.
For Research Use Only
Protein Aliases: ARSA-I; ARSA1; Arsenical pump-driving ATPase; Arsenite-stimulated ATPase; ATPase GET3; Guided entry of tail-anchored proteins factor 3, ATPase; hARSA-I; hASNA-I; MGC3821; Transmembrane domain recognition complex 40 kDa ATPase subunit
Gene Aliases: ARSA; ARSA-I; ARSA1; ASNA-I; ASNA1; GET3; TRC40
UniProt ID: (Human) O43681
Entrez Gene ID: (Human) 439
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.