Novus Biologicals
Manufacturer Code:NBP297916AF700
Catalog # NB049394
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human ASMTL fragment. |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acetylserotonin N-methyltransferase-like; acetylserotonin N-methyltransferase-like acetylserotonin O-methyltransferase-like ASMTLX ASMTLY ASTML N-acetylserotonin O-methyltransferase-like protein; ASMTL; dTTP/UTP pyrophosphatase; dTTPase/UTPase; N-acetylserotonin O-methyltransferase-like protein; Nucleoside triphosphate pyrophosphatase; Nucleotide PPase; Nucleotide pyrophosphatase; Probable bifunctional dTTP/UTP pyrophosphatase/methyltransferase protein
Gene Aliases: ASMTL; ASMTLX; ASMTLY; ASTML
UniProt ID: (Human) O95671
Entrez Gene ID: (Human) 8623
Molecular Function:
methyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.