Invitrogen
Manufacturer Code:PA577047
Catalog # PIPA577047
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human ASM. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acid sphingomyelinase; ASMase; Sphingomyelin phosphodiesterase; Sphingomyelin phosphodiesterase (EC:3.1.4.12) Acid sphingomyelinase aSMase SMPD1 ASM; sphingomyelin phosphodiesterase 1, acid lysosomal
Gene Aliases: A-SMase; ASM; ASMASE; NPD; SMPD1; Zn-SMase
UniProt ID: (Human) P17405, (Mouse) Q04519
Entrez Gene ID: (Human) 6609, (Rat) 308909, (Mouse) 20597
Molecular Function: hydrolase phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.