Novus Biologicals
Manufacturer Code:NBP267481
Catalog # NBP267481
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SU36-06 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: apoptosis signal regulating kinase 1; Apoptosis signal-regulating kinase 1; Apoptosis signal-regulating kinase 1 ASK-1 ASK1MEKK 5 EC 2.7.11 MAP/ERK kinase kinase 5 MAPK/ERK kinase kinase 5 MAPKKK5EC 2.7.11.25 MEK kinase 5 MEKK5apoptosis signal regulating kinase 1 mitogen-activated protein kinase kinase kinase 5; ASK-1; MAP/ERK kinase kinase 5; MAPK/ERK kinase kinase 5; MEK kinase 5; MEKK 5; Mitogen-activated protein kinase kinase kinase 5
Gene Aliases: ASK1; MAP3K5; MAPKKK5; MEKK5
UniProt ID: (Human) Q99683
Entrez Gene ID: (Human) 4217
Molecular Function: kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.