Invitrogen
Manufacturer Code:PA577052
Catalog # PIPA577052
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human ASIP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: agouti signaling protein, nonagouti homolog; Agouti switch protein; Agouti-signaling protein; Agouti-signaling protein ASP Agouti switch protein ASIP AGTI AGTIL ASP; ASP; nonagouti homolog
Gene Aliases: AGSW; AGTI; AGTIL; ASIP; ASP; SHEP9
UniProt ID: (Human) P42127
Entrez Gene ID: (Human) 434
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.