Novus Biologicals
Manufacturer Code:NBP304874100UL
Catalog # NB093819
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 23-132 of human ASIP (NP_001663.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: agouti (mouse)-signaling protein agouti signaling protein agouti signaling protein nonagouti homolog agouti signaling protein nonagouti homolog (mouse) Agouti switch protein agouti-signaling protein AGTI AGTILMGC126092 ASPAGSW MGC126093 nonagouti homolog SHEP9; agouti signaling protein, nonagouti homolog; Agouti switch protein; Agouti-signaling protein; ASP; nonagouti homolog
Gene Aliases: AGSW; AGTI; AGTIL; ASIP; ASP; SHEP9
UniProt ID: (Human) P42127
Entrez Gene ID: (Human) 434
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.