Invitrogen
Manufacturer Code:PA587795
Catalog # PIPA587795
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 140-330 of human ASIC5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acid-sensing ion channel 5; Amiloride-sensitive cation channel 5; amiloride-sensitive cation channel 5, intestinal; amiloride-sensitive Na+ channel; amiloride-sensitive Na+ channel (BLINaC gene); amiloride-sensitive sodium channel; ASIC5; BLINaC; Brain-liver-intestine amiloride-sensitive Na(+) channel; brain-liver-intestine amiloride-sensitive sodium channel; inactivation no afterpotential C
Gene Aliases: Accn5; Asic5; Blinac; Inac
UniProt ID: (Mouse) Q9R0Y1, (Rat) Q9R0W5
Entrez Gene ID: (Mouse) 58170, (Rat) 63866
Molecular Function: ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.