Invitrogen
Manufacturer Code:PA556148
Catalog # PIPA556148
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ASIC5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acid sensing (proton gated) ion channel family member 5; acid sensing ion channel family member 5; acid-sensing (proton-gated) ion channel family member 5; Acid-sensing ion channel 5; Acid-sensing ion channel 5 ASIC5 Amiloride-sensitive cation channel 5 Human intestine Na(+) channel HINaC ASIC5 ACCN5 HINAC; Amiloride-sensitive cation channel 5; amiloride-sensitive cation channel 5, intestinal; amiloride-sensitive sodium channel; ASIC5; HINaC; Human intestine Na(+) channel
Gene Aliases: ACCN5; ASIC5; HINAC; INAC
UniProt ID: (Human) Q9NY37
Entrez Gene ID: (Human) 51802
Molecular Function: ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.