Invitrogen
Manufacturer Code:PA595378
Catalog # PIPA595378
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the N-terminus of human ACCN1 different from the related mouse and rat sequences by one amino acid. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acid sensing (proton gated) ion channel 2; acid sensing ion channel 2; acid-sensing (proton-gated) ion channel 2; Acid-sensing ion channel 2; Acid-sensing ion channel 2 ASIC2 Amiloride-sensitive brain sodium channel Amiloride-sensitive cation channel 1 neuronal Amiloride-sensitive cation channel neuronal 1 Brain sodium channel 1 BNC1 BNaC1 Mammalian degenerin homolog ASIC2 ACCN ACCN1 BNAC1 MDEG; Amiloride-sensitive brain sodium channel; Amiloride-sensitive brain sodium channel 2; Amiloride-sensitive cation channel 1, neuronal; Amiloride-sensitive cation channel neuronal 1; ASIC2; BNC1; Brain sodium channel 1; degenerin; Mammalian degenerin homolog; MDEG; neuronal amiloride-sensitive cation channel 1
Gene Aliases: ACCN; ACCN1; ASIC2; ASIC2a; BNAC1; BNC1; BNC1k; hBNaC1; MDEG; MDEG1; MDEG2
UniProt ID: (Human) Q16515, (Rat) Q62962
Entrez Gene ID: (Human) 40, (Rat) 25364
Molecular Function: ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.