Proteintech
Manufacturer Code:178511AP150UL
Catalog # 50-173-1832
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ASIC2 Fusion Protein Ag12301 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACCN ACCN1 Acid sensing ion channel 2 ASIC2 ASIC2a BNaC1 BNC1 Brain sodium channel 1 hBNaC1 Mammalian degenerin homolog MDEG; acid sensing (proton gated) ion channel 2; acid sensing ion channel 2; acid-sensing (proton-gated) ion channel 2; Acid-sensing ion channel 2; Amiloride-sensitive brain sodium channel; Amiloride-sensitive brain sodium channel 2; Amiloride-sensitive cation channel 1, neuronal; amiloride-sensitive cation channel 1, neuronal (degenerin); Amiloride-sensitive cation channel neuronal 1; ASIC2; BNC1; Brain sodium channel 1; degenerin; Mammalian degenerin homolog; MDEG; neuronal amiloride-sensitive cation channel 1
Gene Aliases: ACCN; ACCN1; ACIC2; ASIC2; ASIC2a; BNAC1; BNaC1a; BNC1; BNC1k; hBNaC1; MDEG; MDEG1; MDEG2
UniProt ID: (Human) Q16515, (Rat) Q62962, (Mouse) Q925H0
Entrez Gene ID: (Human) 40, (Rat) 25364, (Mouse) 11418
Molecular Function: ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.