Invitrogen
Manufacturer Code:MA527640
Catalog # PIMA527640
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S271-44 |
Immunogen | Fusion protein amino acids 460-526 (Cytoplasmic C-terminus) of mouse ASIC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ACCN2 Acid-sensing (proton-gated) ion channel Acid-sensing ion channel 1 Amiloride-sensitive cation channel 2 neuronal BNAC2 Brain sodium channel 2; acid sensing ion channel; Acid-sensing ion channel; Acid-sensing ion channel 1; Amiloride-sensitive cation channel 2, neuronal; ASIC1; BNaC2; Brain sodium channel 2; degenerin 2; proton gated cation channel ASIC1
Gene Aliases: Accn2; AI843610; ASIC; Asic1; ASIC1a; B530003N02Rik; BNaC2
UniProt ID: (Mouse) Q6NXK8, (Rat) P55926
Entrez Gene ID: (Mouse) 11419, (Rat) 79123
Molecular Function: ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.