OriGene
Manufacturer Code:TA810986
Catalog # 50-168-3750
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI10D8 |
Immunogen | Human recombit protein fragment corresponding to amino acids 347-628 of human ASH2L produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ash2 (absent, small, or homeotic)-like; ASH2 ASH2L1 ASH2L2 Bre2; ASH2-like protein; Set1/Ash2 histone methyltransferase complex subunit ASH2
Gene Aliases: ASH2; ASH2L; ASH2L1; ASH2L2; Bre2
UniProt ID: (Human) Q9UBL3
Entrez Gene ID: (Human) 9070
Molecular Function:
DNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.