Invitrogen
Manufacturer Code:PA540842
Catalog # PIPA540842
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human ASH2L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ash2 (absent small or homeotic)-like (Drosophila) ASH2 ASH2L1 ASH2L2 Bre2; ash2 (absent, small, or homeotic)-like; ASH2-like protein; Set1/Ash2 histone methyltransferase complex subunit ASH2
Gene Aliases: ASH2; ash2-like; ASH2L; ASH2L1; ASH2L2; Bre2
UniProt ID: (Human) Q9UBL3, (Mouse) Q91X20
Entrez Gene ID: (Rabbit) 100338600, (Human) 9070, (Bovine) 100138350, (Horse) 100059465, (Dog) 475589, (Guinea pig) 100732899, (Mouse) 23808, (Rat) 290829
Molecular Function: DNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.