Novus Biologicals
Manufacturer Code:NB600250
Catalog # NB600250
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 50 and 100 of human Absent Small or Homeotic-Like 2 using the numbering given in entry NP_004665.1 (GeneID 9070). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ash2 (absent, small, or homeotic)-like; ASH2 ash2 (absent small or homeotic)-like (Drosophila) ASH2L1ash2 (absent small or homeotic Drosophila homolog)-like ASH2L2ASH2-like protein Bre2 set1/Ash2 histone methyltransferase complex subunit ASH2; ASH2-like protein; Set1/Ash2 histone methyltransferase complex subunit ASH2
Gene Aliases: ASH2; ASH2L; ASH2L1; ASH2L2; Bre2
UniProt ID: (Human) Q9UBL3
Entrez Gene ID: (Human) 9070
Molecular Function:
DNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.