Abnova
Manufacturer Code:PAB31297
Catalog # 89-998-412
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to human ASH2L. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ASH2; ash2 (absent, small, or homeotic)-like; ASH2-like protein; ASH2L1; ASH2L2; Bre2; Set1/Ash2 histone methyltransferase complex subunit ASH2
Gene Aliases: ASH2; ASH2L; ASH2L1; ASH2L2; Bre2
UniProt ID: (Human) Q9UBL3
Entrez Gene ID: (Human) 9070
Molecular Function: DNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.