Novus Biologicals
Manufacturer Code:NB10093289
Catalog # NB10093289
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 1925 and 1975 of human absent small and homeotic disks protein 1 using the numbering given in entry CAI12722.1 (GeneID 55870). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Absent small and homeotic disks protein 1 homolog; Absent small and homeotic disks protein 1 homolog ASH1 ash1 (absent small or homeotic)-like (Drosophila) ASH1L1 ASH1-like protein EC 2.1.1.43 FLJ10504 huASH1 KMT2HKIAA1420 Lysine N-methyltransferase 2H probable histone-lysine N-methyltransferase ASH1L; ash1 (absent, small, or homeotic)-like; ASH1-like protein; Histone-lysine N-methyltransferase ASH1L; huASH1; Lysine N-methyltransferase 2H; probable histone-lysine N-methyltransferase ASH1L
Gene Aliases: ASH1; ASH1L; ASH1L1; KIAA1420; KMT2H
UniProt ID: (Human) Q9NR48
Entrez Gene ID: (Human) 55870
Molecular Function:
DNA binding protein
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.