Proteintech
Manufacturer Code:668311IG150UL
Catalog # 50-173-7219
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2A5C12 |
Immunogen | ASGR2 Fusion Protein Ag2037 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ASGP R ASGP R 2 ASGPR 2 ASGR2 asialoglycoprotein receptor 2 CLEC4H2 Hepatic lectin H2 HL 2 Hs.1259 L H2; ASGP-R 2; Asialoglycoprotein receptor 2; C-type lectin domain family 4 member H2; HBxAg-binding protein; Hepatic lectin H2; HL-2
Gene Aliases: ASGP-R2; ASGPR2; ASGR2; CLEC4H2; HBXBP; HL-2
UniProt ID: (Human) P07307
Entrez Gene ID: (Human) 433
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.