Abnova
Manufacturer Code:H00000433M05
Catalog # 89-016-225
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1D7 |
Immunogen | ASGR2 (AAH17251 1 a.a. ~ 287 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant ASGR2.
For Research Use Only
Protein Aliases: ASGP-R; ASGP-R 2; Asialoglycoprotein receptor 2; C-type lectin domain family 4 member H2; CLEC4H2; HBxAg-binding protein; Hepatic lectin H2; HL-2; Hs.1259; L-H2
Gene Aliases: ASGP-R2; ASGPR2; ASGR2; CLEC4H2; HBXBP; HL-2
UniProt ID: (Human) P07307
Entrez Gene ID: (Human) 433
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.