Invitrogen
Manufacturer Code:PA540783
Catalog # PIPA540783
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human ASGR2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ankyrin repeat domain 12 ANCO1 GAC-1 ANCO-2 Nbla00144 ASGR2; ASGP-R 2; ASGPR 2; Asialoglycoprotein receptor 2; C-type lectin domain family 4 member H2; HBxAg-binding protein; hepatic lectin 2 RHL2; hepatic lectin 2, RHL2; Hepatic lectin H2; Hepatic lectin R2/3; HL-2; RHL-2
Gene Aliases: ASGP-R2; ASGPR2; Asgr-2; ASGR2; CLEC4H2; HBXBP; HL-2
UniProt ID: (Human) P07307, (Rat) P08290
Entrez Gene ID: (Pig) 100739379, (Human) 433, (Rat) 29403
Molecular Function: cell adhesion molecule cytokine receptor defense/immunity protein immunoglobulin receptor superfamily receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.