Invitrogen
Manufacturer Code:MA529045
Catalog # PIMA529045
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 21 |
Immunogen | Recombit Human ASGPR1 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: ASGP-R 1; ASGR1; Asialoglycoprotein receptor 1; C-type lectin domain family 4 member H1; Hepatic lectin H1; HL-1
Gene Aliases: ASGPR; ASGPR1; ASGR1; CLEC4H1; HL-1
UniProt ID: (Human) P07306
Entrez Gene ID: (Human) 432
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.