Invitrogen
Manufacturer Code:MA529654
Catalog # PIMA529654
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 114 |
Immunogen | Recombit Mouse ASGPR1/ASGR1 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: ASGP-R 1; ASGPR 1; ASGR1; Asialoglycoprotein receptor 1; Hepatic lectin 1; HL-1
Gene Aliases: ASGPR1; Asgr; Asgr-1; Asgr1; HL-1
UniProt ID: (Mouse) P34927
Entrez Gene ID: (Mouse) 11889
Molecular Function: cell adhesion molecule cytokine receptor defense/immunity protein immunoglobulin receptor superfamily receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.