Invitrogen
Manufacturer Code:PA568619
Catalog # PIPA568619
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N terminal region of human ASGR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ASGP-R 1; ASGPR 1; ASGPR ASGPR1 CLEC4H1 HL-1 C-type lectin domain family 4 member H1 hepatic lectin H1; asialoglycoprotein receptor (RHL1); Asialoglycoprotein receptor 1; Asialoglycoprotein receptor 1 (hepatic lectin); asialoglycoprotein receptor RHL1; C-type lectin domain family 4 member H1; Hepatic lectin 1; hepatic lectin 1 RHL1; hepatic lectin 1, RHL1; Hepatic lectin H1; HL-1; RHL-1
Gene Aliases: ASGPR; ASGPR1; ASGR; Asgr-1; ASGR1; CLEC4H1; HL-1; RATRHL1; RHL1
UniProt ID: (Human) P07306, (Rat) P02706, (Mouse) P34927
Entrez Gene ID: (Bovine) 509121, (Pig) 100625649, (Human) 432, (Horse) 100061471, (Rat) 24210, (Mouse) 11889, (Guinea pig) 100714188
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.