Invitrogen
Manufacturer Code:PA532030
Catalog # PIPA532030
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 25 and 291 of Human ASGPR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ASGP-R 1; ASGPR 1; ASGPR ASGPR1 CLEC4H1 HL-1 C-type lectin domain family 4 member H1 hepatic lectin H1; Asialoglycoprotein receptor 1; C-type lectin domain family 4 member H1; Hepatic lectin 1; Hepatic lectin H1; HL-1
Gene Aliases: ASGPR; ASGPR1; Asgr; Asgr-1; ASGR1; CLEC4H1; HL-1
UniProt ID: (Human) P07306, (Mouse) P34927
Entrez Gene ID: (Human) 432, (Mouse) 11889
Molecular Function: cell adhesion molecule cytokine receptor defense/immunity protein immunoglobulin receptor superfamily receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.