Abnova
Manufacturer Code:DH0051
Catalog # 89-339-152
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Immunogen | Human recombinant ASGPR1 |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 2 to 8°C. Do not freeze. |
Mouse Monoclonal Antibody.
Protein Aliases: ASGP-R 1; Asialoglycoprotein receptor 1; C-type lectin domain family 4 member H1; Hepatic lectin H1; HL-1; HL-1 ASGPR ASGPR1 CLEC4H1
Gene Aliases: ASGPR; ASGPR1; ASGR1; CLEC4H1; HL-1
UniProt ID: (Human) P07306
Entrez Gene ID: (Human) 432
Molecular Function: cell adhesion molecule cytokine receptor defense/immunity protein immunoglobulin receptor superfamily receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.