Novus Biologicals
Manufacturer Code:NBP250108G
Catalog # NBP250108G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | ASF 4A1/3 |
Immunogen | FLAG-ASF1A |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Anti-silencing function protein 1 homolog A; ASF1 anti-silencing function 1 homolog A; ASF1 silencing function 1 homolog A (S. cerevisiae) CCG1-interacting factor A CIACGI-98 DKFZP547E2110 hAsf1 hAsf1a hCIA histone chaperone ASF1A HSPC146 silencing function 1A silencing function protein 1 homolog A; CCG1-interacting factor A; CIA; hAsf1; hAsf1a; hCIA; Histone chaperone ASF1A
Gene Aliases: ASF1A; CGI-98; CIA; HSPC146
UniProt ID: (Human) Q9Y294
Entrez Gene ID: (Human) 25842
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.