Novus Biologicals
Manufacturer Code:NBP261683
Catalog # NBP2-61683
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2A5F8 |
Immunogen | Purified recombinant fragment of human ASF1A (AA: 1-204) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: Anti-silencing function protein 1 homolog A; ASF1 anti-silencing function 1 homolog A; ASF1 silencing function 1 homolog A (S. cerevisiae) CCG1-interacting factor A CIACGI-98 DKFZP547E2110 hAsf1 hAsf1a hCIA histone chaperone ASF1A HSPC146 silencing function 1A silencing function protein 1 homolog A; CCG1-interacting factor A; CIA; hAsf1; hAsf1a; hCIA; Histone chaperone ASF1A
Gene Aliases: ASF1A; CGI-98; CIA; HSPC146
UniProt ID: (Human) Q9Y294
Entrez Gene ID: (Human) 25842
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.