Invitrogen
Manufacturer Code:PA534201
Catalog # PIPA534201
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 15 amino acid peptide from C-Terminus of human SLC1A5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATB(0); Baboon M7 virus receptor; neutral amino acid transporter B; Neutral amino acid transporter B(0); RD114 virus receptor; RD114/simian type D retrovirus receptor; SLC1A5 ATBO ASCT2 ATB0 Baboon M7 virus receptor AAAT M7VS1 M7V1 R16 RDRC RD114 virus receptor RDR ATB(0); Sodium-dependent neutral amino acid transporter type 2; solute carrier family 1 (neutral amino acid transporter), member 5; Solute carrier family 1 member 5
Gene Aliases: AAAT; ASCT2; ATBO; M7V1; M7VS1; R16; RDR; RDRC; SLC1A5
UniProt ID: (Human) Q15758
Entrez Gene ID: (Human) 6510
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.