Novus Biologicals
Manufacturer Code:NBP2839400.1ML
Catalog # NB031869
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human ASCL2/Mash2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: achaete-scute complex (Drosophila) homolog-like 2 achaete-scute complex homolog 2 (Drosophila) achaete-scute complex-like 2 (Drosophila) ASH2 ASH-2 BHLHA45 bHLHa45achaete-scute homolog 2 Class A basic helix-loop-helix protein 45 hASH2 HASH2achaete-scute complex-like 2 mammalian achaete/scute homologue 2 MASH2; achaete-scute complex homolog 2; achaete-scute complex-like 2; Achaete-scute homolog 2; ASH-2; bHLHa45; Class A basic helix-loop-helix protein 45; mammalian achaete/scute homologue 2; Mash2
Gene Aliases: ASCL2; ASH2; BHLHA45; HASH2; MASH2
UniProt ID: (Human) Q99929
Entrez Gene ID: (Human) 430
Molecular Function:
basic helix-loop-helix transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.