R&D Systems
Manufacturer Code:AF2567
Catalog # AF2567
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant mouse ASCL1/Mash1 His74-Asp173 (Gln160Glu) Accession # Q53WV9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: achaete scute protein; achaete-scute complex (Drosophila) homolog-like 1 achaete-scute complex homolog 1 (Drosophila) achaete-scute complex-like 1 (Drosophila) achaete-scute homolog 1 ASCL1 ASH1 ASH-1 ASH1MASH1 BHLHA46 bHLHa46achaete-scute complex-like 1 Class A basic helix-loop-helix protein 46 HASH1 HASH1achaete scute protein Mash1; achaete-scute complex homolog 1; achaete-scute complex-like 1; Achaete-scute homolog 1; ASH-1; bHLHa46; Class A basic helix-loop-helix protein 46
Gene Aliases: ASCL1; ASH1; BHLHA46; HASH1; MASH1
UniProt ID: (Human) P50553
Entrez Gene ID: (Human) 429
Molecular Function:
basic helix-loop-helix transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.