Invitrogen
Manufacturer Code:PA586633
Catalog # PIPA586633
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 140-190 of Human ASCL1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: achaete scute protein; achaete-scute complex homolog 1; achaete-scute complex homolog-like 1; achaete-scute complex-like 1; achaete-scute complex-like protein 1; Achaete-scute homolog 1; Achaete-scute homolog 1 ASH-1 hASH1 Class A basic helix-loop-helix protein 46 bHLHa46 ASCL1 ASH1 BHLHA46 HASH1 Mash-1 Mash1; ASH-1; bHLHa46; Class A basic helix-loop-helix protein 46; mammalian achaete scute homolog 1; mASH-1
Gene Aliases: AI225900; ASCL1; ASH1; BHLHA46; HASH1; Mash-1; MASH1
UniProt ID: (Human) P50553, (Rat) P19359, (Mouse) Q02067
Entrez Gene ID: (Human) 429, (Rat) 64186, (Mouse) 17172
Molecular Function:
basic helix-loop-helix transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.