Invitrogen
Manufacturer Code:PA547348
Catalog # PIPA547348
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit mouse ASCL1/Mash1 His74-Asp173 (Gln160Glu) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: achaete-scute complex homolog-like 1; achaete-scute complex-like protein 1; Achaete-scute homolog 1; Achaete-scute homolog 1 ASH-1 hASH1 Class A basic helix-loop-helix protein 46 bHLHa46 ASCL1 ASH1 BHLHA46 HASH1 Mash-1 Mash1; ASH-1; mammalian achaete scute homolog 1; mASH-1
Gene Aliases: AI225900; Ascl1; ASH1; bHLHa46; Mash-1; Mash1
UniProt ID: (Mouse) Q02067
Entrez Gene ID: (Mouse) 17172
Molecular Function:
basic helix-loop-helix transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.