Abnova
Manufacturer Code:PAB1911
Catalog # 89-120-641
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human ASCL1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of ASCL1.
For Research Use Only
Protein Aliases: achaete scute protein; achaete-scute complex homolog 1; achaete-scute complex-like 1; Achaete-scute homolog 1; ASH-1; ASH1; bHLHa46; Class A basic helix-loop-helix protein 46; HASH1; MASH1
Gene Aliases: ASCL1; ASH1; BHLHA46; HASH1; MASH1
UniProt ID: (Human) P50553
Entrez Gene ID: (Human) 429
Molecular Function:
basic helix-loop-helix transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.