Novus Biologicals
Manufacturer Code:NBP203818
Catalog # NBP203818
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3E9 |
Immunogen | Full length human recombinant protein of human PYCARD (NP_037390) produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Apoptosis-associated speck-like protein containing a CARD; ASCMGC10332 CARD5TMS caspase recruitment domain protein 5 Caspase recruitment domain-containing protein 5 hASC PYD and CARD domain containing PYD and CARD domain-containing protein Target of methylation-induced silencing 1 TMS-1 TMS1apoptosis-associated speck-like protein containing a CARD; Caspase recruitment domain-containing protein 5; hASC; PYD and CARD domain-containing protein; Target of methylation-induced silencing 1
Gene Aliases: ASC; CARD5; PYCARD; TMS; TMS-1; TMS1
UniProt ID: (Human) Q9ULZ3
Entrez Gene ID: (Human) 29108
Molecular Function:
cysteine protease
enzyme modulator
hydrolase
protease
protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.