Invitrogen
Manufacturer Code:PA575603
Catalog # PIPA575603
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues in Human ACER1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acylsphingosine deacylase 3; alkaline CDase 1; Alkaline ceramidase 1; Alkaline ceramidase 1 AlkCDase 1 Alkaline CDase 1 EC&equals3.5.1.23 Acylsphingosine deacylase 3 N-acylsphingosine amidohydrolase 3 ACER1 ASAH3; AlkCDase 1; CTB-180A7.3; maCER1; N-acylsphingosine amidohydrolase (alkaline ceramidase) 3; N-acylsphingosine amidohydrolase 3
Gene Aliases: 2310024P18Rik; ACER1; AI662009; ALKCDase1; ASAH3; Cer1
UniProt ID: (Human) Q8TDN7, (Mouse) Q8R4X1
Entrez Gene ID: (Human) 125981, (Rat) 301118, (Mouse) 171168
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.