Novus Biologicals
Manufacturer Code:NBP174125
Catalog # NBP174125
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to the C terminal of ACER1. Immunizing peptide sequence ITFPYGMVTMALVDANYEMPGETLKVRYWPRDSWPVGLPYVEIRGDDKDC. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acylsphingosine deacylase 3; Acylsphingosine deacylase 3 Alkaline CDase 1 alkaline ceramidase 1 AlkCDase 1 ALKCDase1 ASAH3 EC 3.5.1.23 MGC138327 MGC138329 N-acylsphingosine amidohydrolase (alkaline ceramidase) 3 N-acylsphingosine amidohydrolase 3; alkaline CDase 1; Alkaline ceramidase 1; AlkCDase 1; CTB-180A7.3; N-acylsphingosine amidohydrolase (alkaline ceramidase) 3; N-acylsphingosine amidohydrolase 3
Gene Aliases: ACER1; ALKCDase1; ASAH3
UniProt ID: (Human) Q8TDN7
Entrez Gene ID: (Human) 125981
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.