Novus Biologicals
Manufacturer Code:NBP298204R
Catalog # NB081313
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant ASAH2/N-acylsphingosine Amidohydrolase-2 (NP_061300.1). |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acylsphingosine deacylase 2; ASAH2 N-acylsphingosine amidohydrolase (non-lysosomal ceramidase) 2 BCDase HNAC1 LCDase NCDase N-CDase; BCDase; hCD; LCDase; mitochondrial ceramidase; N-acylsphingosine amidohydrolase (non-lysosomal ceramidase) 2; N-acylsphingosine amidohydrolase 2; N-CDase; Neutral ceramidase; Neutral ceramidase soluble form; neutral/alkaline ceramidase; Non-lysosomal ceramidase
Gene Aliases: ASAH2; BCDase; HNAC1; LCDase; N-CDase; NCDase
UniProt ID: (Human) Q9NR71
Entrez Gene ID: (Human) 56624
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.