Invitrogen
Manufacturer Code:MA531037
Catalog # PIMA531037
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 2 |
Immunogen | Recombit Mouse ASAH2 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: Acylsphingosine deacylase 2; BCDase HNAC1 LCDase N-CDase NCDase N-acylsphingosine amidohydrolase 2 acylsphingosine deacylase 2 mitochondrial ceramidase neutral ceramidase neutral/alkaline ceramidase non-lysosomal ceramidase; N-acylsphingosine amidohydrolase 2; N-CDase; NCDase; Neutral ceramidase; Neutral ceramidase soluble form; neutral/alkaline ceramidase
Gene Aliases: AI585898; Asah2
UniProt ID: (Mouse) Q9JHE3
Entrez Gene ID: (Mouse) 54447
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.